Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611472 MIM: 603056 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MBD5 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
MBD5 - methyl-CpG binding domain protein 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018328.4 | Intron | NP_060798.2 | ||||
XM_011511470.2 | Intron | XP_011509772.1 | ||||
XM_011511472.2 | Intron | XP_011509774.1 | ||||
XM_017004482.1 | Intron | XP_016859971.1 | ||||
XM_017004483.1 | Intron | XP_016859972.1 | ||||
XM_017004484.1 | Intron | XP_016859973.1 | ||||
XM_017004485.1 | Intron | XP_016859974.1 |
ORC4 - origin recognition complex subunit 4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |