Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603785 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MPDZ PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MPDZ - multiple PDZ domain crumbs cell polarity complex component | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001261406.1 | 6582 | UTR 3 | NP_001248335.1 | |||
NM_001261407.1 | 6582 | UTR 3 | NP_001248336.1 | |||
NM_003829.4 | 6582 | UTR 3 | NP_003820.2 | |||
XM_005251622.4 | 6582 | UTR 3 | XP_005251679.1 | |||
XM_005251623.4 | 6582 | UTR 3 | XP_005251680.1 | |||
XM_006716885.3 | 6582 | UTR 3 | XP_006716948.1 | |||
XM_006716886.3 | 6582 | UTR 3 | XP_006716949.1 | |||
XM_006716887.3 | 6582 | UTR 3 | XP_006716950.1 | |||
XM_006716888.3 | 6582 | UTR 3 | XP_006716951.1 | |||
XM_006716889.3 | 6582 | UTR 3 | XP_006716952.1 | |||
XM_006716891.3 | 6582 | UTR 3 | XP_006716954.1 | |||
XM_017015252.1 | 6582 | UTR 3 | XP_016870741.1 | |||
XM_017015253.1 | 6582 | UTR 3 | XP_016870742.1 | |||
XM_017015254.1 | 6582 | UTR 3 | XP_016870743.1 | |||
XM_017015255.1 | 6582 | UTR 3 | XP_016870744.1 | |||
XM_017015256.1 | 6582 | UTR 3 | XP_016870745.1 | |||
XM_017015257.1 | 6582 | UTR 3 | XP_016870746.1 | |||
XM_017015258.1 | 6582 | Intron | XP_016870747.1 | |||
XM_017015259.1 | 6582 | Intron | XP_016870748.1 |