Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605238 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
HNMT PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
HNMT - histamine N-methyltransferase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001024074.2 | 727 | UTR 3 | NP_001019245.1 | |||
NM_001024075.1 | 727 | Intron | NP_001019246.1 | |||
NM_006895.2 | 727 | Intron | NP_008826.1 | |||
XM_011511064.2 | 727 | Intron | XP_011509366.1 | |||
XM_017003948.1 | 727 | Intron | XP_016859437.1 | |||
XM_017003949.1 | 727 | Intron | XP_016859438.1 |