Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 601991 | ||||||||||||||||||||
Literature Links: |
NOVA2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
NOVA2 - NOVA alternative splicing regulator 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002516.3 | 1057 | Silent Mutation | GCA,GCG | A,A 319 | NP_002507.1 | |
XM_006723230.3 | 1057 | Silent Mutation | GCA,GCG | A,A 210 | XP_006723293.1 | |
XM_017026838.1 | 1057 | Silent Mutation | GCA,GCG | A,A 210 | XP_016882327.1 | |
XM_017026839.1 | 1057 | Silent Mutation | GCA,GCG | A,A 210 | XP_016882328.1 | |
XM_017026840.1 | 1057 | Silent Mutation | GCA,GCG | A,A 210 | XP_016882329.1 |