Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612323 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
IMMP1L PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU) - Not Available | |||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese)
|
|||||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||||||||
EUR
|
|||||||||||
AMR
|
IMMP1L - inner mitochondrial membrane peptidase subunit 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001304274.1 | Intron | NP_001291203.1 | ||||
NM_144981.2 | Intron | NP_659418.1 | ||||
XM_005252812.3 | Intron | XP_005252869.1 | ||||
XM_011519942.1 | Intron | XP_011518244.1 | ||||
XM_011519943.2 | Intron | XP_011518245.1 | ||||
XM_011519944.2 | Intron | XP_011518246.1 | ||||
XM_011519945.2 | Intron | XP_011518247.1 | ||||
XM_011519946.2 | Intron | XP_011518248.1 | ||||
XM_011519947.2 | Intron | XP_011518249.1 | ||||
XM_017017305.1 | Intron | XP_016872794.1 | ||||
XM_017017306.1 | Intron | XP_016872795.1 | ||||
XM_017017307.1 | Intron | XP_016872796.1 |
Set Membership: |
HapMap Validated |