Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
1 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 151525 MIM: 615354 | ||||||||||||||||||||
Literature Links: |
CD53 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CD53 - CD53 molecule | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000560.3 | 2542 | Intron | NP_000551.1 | |||
NM_001040033.1 | 2542 | Intron | NP_001035122.1 | |||
NM_001320638.1 | 2542 | Intron | NP_001307567.1 |
LRIF1 - ligand dependent nuclear receptor interacting factor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001006945.1 | 2542 | Intron | NP_001006946.1 | |||
NM_018372.3 | 2542 | Intron | NP_060842.3 | |||
XM_005271029.3 | 2542 | Intron | XP_005271086.1 | |||
XM_011541768.1 | 2542 | Intron | XP_011540070.1 | |||
XM_017001768.1 | 2542 | Intron | XP_016857257.1 | |||
XM_017001769.1 | 2542 | UTR 3 | XP_016857258.1 |