Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
37 submissions
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606510 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
FCRL3 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese)
|
||||||
EUR
|
||||||||
AMR
|
FCRL3 - Fc receptor like 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001320333.1 | 2286 | Missense Mutation | CCG,CTG | P,L 660 | NP_001307262.1 | |
NM_052939.3 | 2286 | Missense Mutation | CCG,CTG | P,L 660 | NP_443171.2 | |
XM_006711145.1 | 2286 | Missense Mutation | CCG,CTG | P,L 660 | XP_006711208.1 |
Set Membership: |
HapMap |