Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604482 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
RNU4-1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||||||||
EUR
|
|||||||||||
AMR
|
RNU4-1 - RNA, U4 small nuclear 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RNU4-2 - RNA, U4 small nuclear 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SIRT4 - sirtuin 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_012240.2 | Intron | NP_036372.1 | ||||
XM_005253865.4 | Intron | XP_005253922.1 | ||||
XM_006719308.2 | Intron | XP_006719371.1 | ||||
XM_006719309.3 | Intron | XP_006719372.1 |