Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 300398 MIM: 300680 MIM: 300036 | ||||||||||||||||||||
Literature Links: |
BCAP31 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
BCAP31 - B-cell receptor-associated protein 31 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PNCK - pregnancy up-regulated nonubiquitous CaM kinase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SLC6A8 - solute carrier family 6 member 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001142805.1 | 2479 | Intron | NP_001136277.1 | |||
NM_001142806.1 | 2479 | UTR 3 | NP_001136278.1 | |||
NM_005629.3 | 2479 | Intron | NP_005620.1 |