Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGCTCTTGAGGACCTGCCAAATGG[A/C]AGAAGGACAGAGACCTGGAGCCCTA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611736 | |||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
GREB1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
|||||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
GREB1 - growth regulation by estrogen in breast cancer 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014668.3 | 54 | UTR 5 | NP_055483.2 | |||
NM_033090.2 | 54 | Intron | NP_149081.1 | |||
NM_148903.2 | 54 | Intron | NP_683701.2 | |||
XM_005246192.4 | 54 | Intron | XP_005246249.1 | |||
XM_005246196.3 | 54 | Intron | XP_005246253.1 | |||
XM_011510418.2 | 54 | Intron | XP_011508720.1 | |||
XM_011510419.2 | 54 | Intron | XP_011508721.1 | |||
XM_011510422.2 | 54 | Intron | XP_011508724.1 | |||
XM_011510423.2 | 54 | Intron | XP_011508725.1 |
MIR4429 - microRNA 4429 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |