Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACCCTTGAAGGAATGCTACAGCCT[C/G]TTTCTTACCCATTCAAACCAGTCGC
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608254 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PAXIP1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
PAXIP1 - PAX interacting protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_007349.3 | Intron | NP_031375.3 | ||||
XM_005249539.1 | Intron | XP_005249596.1 | ||||
XM_011515982.2 | Intron | XP_011514284.1 | ||||
XM_011515983.1 | Intron | XP_011514285.1 | ||||
XM_017011881.1 | Intron | XP_016867370.1 |
PAXIP1-AS2 - PAXIP1 antisense RNA 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |