Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGAAACCAAGGGTTCCAACATTCAT[T/C]ATCCAGTTTCAGAGACACCCTTGAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608254 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PAXIP1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
PAXIP1 - PAX interacting protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_007349.3 | Intron | NP_031375.3 | ||||
XM_005249539.1 | Intron | XP_005249596.1 | ||||
XM_011515982.2 | Intron | XP_011514284.1 | ||||
XM_011515983.1 | Intron | XP_011514285.1 | ||||
XM_017011881.1 | Intron | XP_016867370.1 |
PAXIP1-AS2 - PAXIP1 antisense RNA 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |