Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608828 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
DROSHA PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
DROSHA - drosha ribonuclease III | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001100412.1 | 4628 | UTR 3 | NP_001093882.1 | |||
NM_013235.4 | 4628 | UTR 3 | NP_037367.3 | |||
XM_005248291.3 | 4628 | UTR 3 | XP_005248348.1 | |||
XM_005248292.3 | 4628 | UTR 3 | XP_005248349.1 | |||
XM_005248293.3 | 4628 | UTR 3 | XP_005248350.1 | |||
XM_005248294.3 | 4628 | UTR 3 | XP_005248351.1 | |||
XM_011514033.2 | 4628 | Intron | XP_011512335.1 | |||
XM_017009399.1 | 4628 | UTR 3 | XP_016864888.1 | |||
XM_017009400.1 | 4628 | UTR 3 | XP_016864889.1 | |||
XM_017009401.1 | 4628 | UTR 3 | XP_016864890.1 | |||
XM_017009402.1 | 4628 | UTR 3 | XP_016864891.1 |
Set Membership: |
HapMap |