Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGCGCCCCATCTTCCTGCCGCCTG[A/C]GGTTGCTGCAGCCTTCTGTCTTCAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
STOX2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
STOX2 - storkhead box 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020225.2 | Intron | NP_064610.1 | ||||
XM_011532129.1 | Intron | XP_011530431.1 | ||||
XM_011532130.1 | Intron | XP_011530432.1 | ||||
XM_011532131.1 | Intron | XP_011530433.1 | ||||
XM_017008466.1 | Intron | XP_016863955.1 | ||||
XM_017008467.1 | Intron | XP_016863956.1 |