Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AACTAGGGTCTCAAAGCTTGAAGAT[A/G]TATCTTCCTTCTGATTGATAAAATA
Species: |
Human | |||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||
Phenotype: |
MIM: 610834 | |||||||||||||||||||||||||||||
Literature Links: |
NAA50 PubMed Links | |||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Japanese
|
CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Chinese
|
JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
NAA50 - N(alpha)-acetyltransferase 50, NatE catalytic subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001308445.1 | Intron | NP_001295374.1 | ||||
NM_025146.3 | Intron | NP_079422.1 |
Set Membership: |
Validated |