Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 604814 | ||||||||||||||||||||
Literature Links: |
LILRA6 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LILRA6 - leukocyte immunoglobulin like receptor A6 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LILRB5 - leukocyte immunoglobulin like receptor B5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001081442.2 | Intron | NP_001074911.1 | ||||
NM_001081443.2 | Intron | NP_001074912.1 | ||||
NM_001304457.1 | Intron | NP_001291386.1 | ||||
NM_006840.4 | Intron | NP_006831.1 | ||||
XM_011526359.2 | Intron | XP_011524661.1 | ||||
XM_011526360.2 | Intron | XP_011524662.1 | ||||
XM_011526361.2 | Intron | XP_011524663.1 | ||||
XM_011526362.2 | Intron | XP_011524664.1 |