Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGAGAAGCTGGTTGGCATTCAGTCC[C/T]GTCTGAGGCTGCATTGTCCAATTGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610837 MIM: 603734 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
BCL2L12 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
BCL2L12 - BCL2 like 12 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
IRF3 - interferon regulatory factor 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001197122.1 | Intron | NP_001184051.1 | ||||
NM_001197123.1 | Intron | NP_001184052.1 | ||||
NM_001197124.1 | Intron | NP_001184053.1 | ||||
NM_001197125.1 | Intron | NP_001184054.1 | ||||
NM_001197126.1 | Intron | NP_001184055.1 | ||||
NM_001197127.1 | Intron | NP_001184056.1 | ||||
NM_001197128.1 | Intron | NP_001184057.1 | ||||
NM_001571.5 | Intron | NP_001562.1 | ||||
XM_006723197.1 | Intron | XP_006723260.1 | ||||
XM_006723198.1 | Intron | XP_006723261.1 | ||||
XM_006723200.1 | Intron | XP_006723263.1 | ||||
XM_006723201.1 | Intron | XP_006723264.1 | ||||
XM_006723202.2 | Intron | XP_006723265.1 | ||||
XM_017026766.1 | Intron | XP_016882255.1 | ||||
XM_017026767.1 | Intron | XP_016882256.1 |
SCAF1 - SR-related CTD associated factor 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |