Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609364 MIM: 609661 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
NLRP2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
NLRP2 - NLR family pyrin domain containing 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001174081.1 | 454 | Silent Mutation | AAA,AAG | K,K 104 | NP_001167552.1 | |
NM_001174082.1 | 454 | Silent Mutation | AAA,AAG | K,K 104 | NP_001167553.1 | |
NM_001174083.1 | 454 | Silent Mutation | AAA,AAG | K,K 81 | NP_001167554.1 | |
NM_017852.3 | 454 | Silent Mutation | AAA,AAG | K,K 104 | NP_060322.1 |
NLRP7 - NLR family pyrin domain containing 7 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |