Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 187430 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CRISP2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CRISP2 - cysteine rich secretory protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001142407.2 | Intron | NP_001135879.1 | ||||
NM_001142408.2 | Intron | NP_001135880.1 | ||||
NM_001142417.2 | Intron | NP_001135889.1 | ||||
NM_001142435.2 | Intron | NP_001135907.1 | ||||
NM_001261822.1 | Intron | NP_001248751.1 | ||||
NM_003296.3 | Intron | NP_003287.1 | ||||
XM_005249349.1 | Intron | XP_005249406.1 | ||||
XM_005249350.1 | Intron | XP_005249407.1 | ||||
XM_005249351.3 | Intron | XP_005249408.1 | ||||
XM_005249352.3 | Intron | XP_005249409.1 | ||||
XM_005249353.3 | Intron | XP_005249410.1 | ||||
XM_005249356.1 | Intron | XP_005249413.1 | ||||
XM_011514841.1 | Intron | XP_011513143.1 | ||||
XM_011514842.1 | Intron | XP_011513144.1 | ||||
XM_011514843.2 | Intron | XP_011513145.1 | ||||
XM_011514844.1 | Intron | XP_011513146.1 |