Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608924 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
FOXP4 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
FOXP4 - forkhead box P4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001012426.1 | 1287 | Intron | NP_001012426.1 | |||
NM_001012427.1 | 1287 | Intron | NP_001012427.1 | |||
NM_138457.2 | 1287 | Intron | NP_612466.1 | |||
XM_006714991.3 | 1287 | Intron | XP_006715054.1 | |||
XM_011514289.2 | 1287 | Intron | XP_011512591.1 | |||
XM_011514290.2 | 1287 | Intron | XP_011512592.1 | |||
XM_011514291.2 | 1287 | Intron | XP_011512593.1 | |||
XM_011514292.2 | 1287 | Intron | XP_011512594.1 | |||
XM_011514293.2 | 1287 | Intron | XP_011512595.1 | |||
XM_017010233.1 | 1287 | UTR 5 | XP_016865722.1 | |||
XM_017010234.1 | 1287 | Intron | XP_016865723.1 |
FOXP4-AS1 - FOXP4 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |