Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603681 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
C2orf70 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C2orf70 - chromosome 2 open reading frame 70 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001105519.2 | Intron | NP_001098989.1 | ||||
NM_001322426.1 | Intron | NP_001309355.1 | ||||
XM_011532810.2 | Intron | XP_011531112.1 | ||||
XM_011532811.2 | Intron | XP_011531113.1 | ||||
XM_017003962.1 | Intron | XP_016859451.1 | ||||
XM_017003963.1 | Intron | XP_016859452.1 | ||||
XM_017003964.1 | Intron | XP_016859453.1 | ||||
XM_017003965.1 | Intron | XP_016859454.1 | ||||
XM_017003966.1 | Intron | XP_016859455.1 |
OTOF - otoferlin | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |