Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AACAGCAGGGGGTTGTGTGTCTGTT[C/T]TGTTTCTCTGCTTGCCGAACTTTCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600291 MIM: 611504 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ADCY3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ADCY3 - adenylate cyclase 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001320613.1 | 4385 | Intron | NP_001307542.1 | |||
NM_004036.4 | 4385 | Intron | NP_004027.2 | |||
XM_006711925.1 | 4385 | Intron | XP_006711988.1 | |||
XM_011532489.1 | 4385 | Intron | XP_011530791.1 | |||
XM_011532490.1 | 4385 | Intron | XP_011530792.1 | |||
XM_011532491.1 | 4385 | Intron | XP_011530793.1 | |||
XM_011532492.2 | 4385 | Intron | XP_011530794.1 | |||
XM_011532493.1 | 4385 | Intron | XP_011530795.1 | |||
XM_011532494.1 | 4385 | Intron | XP_011530796.1 | |||
XM_011532496.1 | 4385 | Intron | XP_011530798.1 | |||
XM_017003186.1 | 4385 | Intron | XP_016858675.1 | |||
XM_017003187.1 | 4385 | Intron | XP_016858676.1 | |||
XM_017003188.1 | 4385 | Intron | XP_016858677.1 | |||
XM_017003189.1 | 4385 | Intron | XP_016858678.1 | |||
XM_017003190.1 | 4385 | Intron | XP_016858679.1 | |||
XM_017003191.1 | 4385 | Intron | XP_016858680.1 | |||
XM_017003192.1 | 4385 | Intron | XP_016858681.1 | |||
XM_017003193.1 | 4385 | Intron | XP_016858682.1 |
CENPO - centromere protein O | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001199803.2 | 4385 | UTR 3 | NP_001186732.1 | |||
NM_001322101.1 | 4385 | UTR 3 | NP_001309030.1 | |||
NM_024322.3 | 4385 | UTR 3 | NP_077298.1 |