Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613527 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
DNAJC27 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
DNAJC27 - DnaJ heat shock protein family (Hsp40) member C27 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001198559.1 | 2188 | UTR 3 | NP_001185488.1 | |||
NM_016544.2 | 2188 | UTR 3 | NP_057628.1 | |||
XM_011532902.1 | 2188 | Intron | XP_011531204.1 | |||
XM_017004269.1 | 2188 | UTR 3 | XP_016859758.1 | |||
XM_017004270.1 | 2188 | UTR 3 | XP_016859759.1 |