Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACACAGCACAGAGGCCAAGCATATG[A/G]CCACTCCAAGGAGGGAGGTTCCACA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602572 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ANXA11 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
ANXA11 - annexin A11 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001157.2 | 1776 | Intron | NP_001148.1 | |||
NM_001278407.1 | 1776 | Intron | NP_001265336.1 | |||
NM_001278408.1 | 1776 | Intron | NP_001265337.1 | |||
NM_001278409.1 | 1776 | Intron | NP_001265338.1 | |||
NM_145868.1 | 1776 | Intron | NP_665875.1 | |||
NM_145869.1 | 1776 | Intron | NP_665876.1 | |||
XM_005269741.4 | 1776 | Intron | XP_005269798.1 | |||
XM_005269742.1 | 1776 | Missense Mutation | GCC,GTC | A,V 512 | XP_005269799.1 | |
XM_006717813.1 | 1776 | Missense Mutation | GCC,GTC | A,V 512 | XP_006717876.1 | |
XM_006717814.3 | 1776 | Missense Mutation | GCC,GTC | A,V 512 | XP_006717877.1 | |
XM_011539735.1 | 1776 | Missense Mutation | GCC,GTC | A,V 512 | XP_011538037.1 | |
XM_011539736.2 | 1776 | Intron | XP_011538038.1 |
Set Membership: |
HapMap |