Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 138320 MIM: 165390 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
GPX1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
GPX1 - glutathione peroxidase 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RHOA - ras homolog family member A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001313941.1 | Intron | NP_001300870.1 | ||||
NM_001313943.1 | Intron | NP_001300872.1 | ||||
NM_001313944.1 | Intron | NP_001300873.1 | ||||
NM_001313945.1 | Intron | NP_001300874.1 | ||||
NM_001313946.1 | Intron | NP_001300875.1 | ||||
NM_001313947.1 | Intron | NP_001300876.1 | ||||
NM_001664.3 | Intron | NP_001655.1 |
Set Membership: |
HapMap |