Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603892 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
EFTUD2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
EFTUD2 - elongation factor Tu GTP binding domain containing 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
HIGD1B - HIG1 hypoxia inducible domain family member 1B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001271880.1 | Intron | NP_001258809.1 | ||||
NM_016438.3 | Intron | NP_057522.1 | ||||
XM_006721946.2 | Intron | XP_006722009.1 | ||||
XM_006721947.3 | Intron | XP_006722010.1 | ||||
XM_006721948.3 | Intron | XP_006722011.1 | ||||
XM_011524891.2 | Intron | XP_011523193.1 | ||||
XM_017024742.1 | Intron | XP_016880231.1 | ||||
XM_017024743.1 | Intron | XP_016880232.1 |