Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614677 MIM: 603892 MIM: 137780 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CCDC103 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CCDC103 - coiled-coil domain containing 103 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
EFTUD2 - elongation factor Tu GTP binding domain containing 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
GFAP - glial fibrillary acidic protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001131019.2 | 2250 | Intron | NP_001124491.1 | |||
NM_001242376.1 | 2250 | Intron | NP_001229305.1 | |||
NM_002055.4 | 2250 | UTR 3 | NP_002046.1 | |||
XM_017024451.1 | 2250 | UTR 3 | XP_016879940.1 |