Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600967 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
C8orf59 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
C8orf59 - chromosome 8 open reading frame 59 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001099670.2 | Intron | NP_001093140.1 | ||||
NM_001099671.2 | Intron | NP_001093141.1 | ||||
NM_001099672.2 | Intron | NP_001093142.1 | ||||
NM_001099673.2 | Intron | NP_001093143.1 | ||||
NM_001293320.1 | Intron | NP_001280249.1 | ||||
XM_011517528.2 | Intron | XP_011515830.1 |
E2F5 - E2F transcription factor 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001083588.1 | Intron | NP_001077057.1 | ||||
NM_001083589.1 | Intron | NP_001077058.1 | ||||
NM_001951.3 | Intron | NP_001942.2 |
Set Membership: |
HapMap |