Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGTCTCACCCAGCTGCTGGAAGAT[A/G]GGGGCTTCATTCCCTAACTAGAGTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603404 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF169 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ZNF169 - zinc finger protein 169 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001301275.1 | Intron | NP_001288204.1 | ||||
NM_003448.2 | Intron | NP_003439.2 | ||||
NM_194320.3 | Intron | NP_919301.2 | ||||
XM_017014362.1 | Intron | XP_016869851.1 | ||||
XM_017014363.1 | Intron | XP_016869852.1 | ||||
XM_017014364.1 | Intron | XP_016869853.1 | ||||
XM_017014365.1 | Intron | XP_016869854.1 | ||||
XM_017014366.1 | Intron | XP_016869855.1 |
Set Membership: |
HapMap |