Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAAGGCCATTGGCCCCAAAGAAAGG[C/T]CTTGTAGGCGACTGTTTAGGGACAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607211 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CARD14 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
CARD14 - caspase recruitment domain family member 14 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001257970.1 | Intron | NP_001244899.1 | ||||
NM_024110.4 | Intron | NP_077015.2 | ||||
NM_052819.2 | Intron | NP_438170.1 | ||||
XM_011525212.1 | Intron | XP_011523514.1 | ||||
XM_011525213.1 | Intron | XP_011523515.1 | ||||
XM_011525215.1 | Intron | XP_011523517.1 | ||||
XM_011525216.1 | Intron | XP_011523518.1 | ||||
XM_011525217.1 | Intron | XP_011523519.1 | ||||
XM_011525218.2 | Intron | XP_011523520.1 | ||||
XM_017025057.1 | Intron | XP_016880546.1 | ||||
XM_017025058.1 | Intron | XP_016880547.1 |