Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613799 MIM: 616637 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CCDC40 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU) - Not Available | ||||||
EAS
|
African American
|
YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
CCDC40 - coiled-coil domain containing 40 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001243342.1 | 89 | Intron | NP_001230271.1 | |||
NM_017950.3 | 89 | Intron | NP_060420.2 | |||
XM_005257492.4 | 89 | Intron | XP_005257549.1 | |||
XM_011524963.2 | 89 | UTR 5 | XP_011523265.1 | |||
XM_011524964.2 | 89 | Intron | XP_011523266.1 | |||
XM_011524965.2 | 89 | Intron | XP_011523267.1 | |||
XM_017024807.1 | 89 | Intron | XP_016880296.1 |
TBC1D16 - TBC1 domain family member 16 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001271844.1 | 89 | Intron | NP_001258773.1 | |||
NM_001271845.1 | 89 | Intron | NP_001258774.1 | |||
NM_001271846.1 | 89 | Intron | NP_001258775.1 | |||
NM_019020.3 | 89 | Intron | NP_061893.2 | |||
XM_005257049.2 | 89 | Intron | XP_005257106.1 | |||
XM_005257050.3 | 89 | Intron | XP_005257107.1 | |||
XM_005257052.3 | 89 | Intron | XP_005257109.1 | |||
XM_006721694.3 | 89 | Intron | XP_006721757.1 | |||
XM_006721695.3 | 89 | Intron | XP_006721758.1 | |||
XM_011524322.2 | 89 | Intron | XP_011522624.1 | |||
XM_011524323.2 | 89 | Intron | XP_011522625.2 | |||
XM_011524324.2 | 89 | Intron | XP_011522626.1 | |||
XM_017024185.1 | 89 | Intron | XP_016879674.1 | |||
XM_017024186.1 | 89 | Intron | XP_016879675.1 | |||
XM_017024187.1 | 89 | Intron | XP_016879676.1 | |||
XM_017024188.1 | 89 | Intron | XP_016879677.1 |
Set Membership: |
Validated |