Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603781 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
RECQL5 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
RECQL5 - RecQ like helicase 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001003715.3 | 1397 | Intron | NP_001003715.1 | |||
NM_001003716.3 | 1397 | Intron | NP_001003716.1 | |||
NM_004259.6 | 1397 | Intron | NP_004250.4 | |||
XM_005257818.3 | 1397 | Intron | XP_005257875.1 | |||
XM_005257822.3 | 1397 | Intron | XP_005257879.1 | |||
XM_005257823.3 | 1397 | Intron | XP_005257880.1 | |||
XM_006722186.2 | 1397 | Intron | XP_006722249.1 | |||
XM_011525482.2 | 1397 | Intron | XP_011523784.1 | |||
XM_011525484.1 | 1397 | Intron | XP_011523786.1 | |||
XM_011525485.2 | 1397 | Intron | XP_011523787.1 | |||
XM_011525486.2 | 1397 | Intron | XP_011523788.1 | |||
XM_017025343.1 | 1397 | Intron | XP_016880832.1 | |||
XM_017025344.1 | 1397 | Intron | XP_016880833.1 |
SMIM5 - small integral membrane protein 5 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SMIM6 - small integral membrane protein 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001162997.1 | 1397 | Intron | NP_001156469.1 | |||
XM_005256903.4 | 1397 | UTR 5 | XP_005256960.1 |
Set Membership: |
HapMap Validated |