Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 186854 | |||||||||||||||||||||||
Literature Links: |
SLC6A6 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
SLC6A6 - solute carrier family 6 member 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001134367.3 | Intron | NP_001127839.2 | ||||
NM_001134368.3 | Intron | NP_001127840.1 | ||||
NM_003043.5 | Intron | NP_003034.2 | ||||
XM_006713307.2 | Intron | XP_006713370.1 | ||||
XM_011534030.2 | Intron | XP_011532332.1 | ||||
XM_011534031.2 | Intron | XP_011532333.1 |