Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAATGAGGGCCTCCCTCCCCAATCC[C/T]GTATGCCATACTCCCTTTAGGCCCA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
25 submissions
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602201 MIM: 616092 MIM: 612805 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ECM1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese)
|
||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
ECM1 - extracellular matrix protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001202858.1 | Intron | NP_001189787.1 | ||||
NM_004425.3 | Intron | NP_004416.2 | ||||
NM_022664.2 | Intron | NP_073155.2 |
FALEC - focally amplified long non-coding RNA in epithelial cancer | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TARS2 - threonyl-tRNA synthetase 2, mitochondrial (putative) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |