Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607521 MIM: 104750 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
HPS5 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
HPS5 - HPS5, biogenesis of lysosomal organelles complex 2 subunit 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_007216.3 | 3799 | UTR 3 | NP_009147.3 | |||
NM_181507.1 | 3799 | UTR 3 | NP_852608.1 | |||
NM_181508.1 | 3799 | UTR 3 | NP_852609.1 | |||
XM_011519862.1 | 3799 | UTR 3 | XP_011518164.1 | |||
XM_011519863.1 | 3799 | UTR 3 | XP_011518165.1 | |||
XM_011519864.1 | 3799 | UTR 3 | XP_011518166.1 | |||
XM_011519865.1 | 3799 | UTR 3 | XP_011518167.1 | |||
XM_011519866.1 | 3799 | UTR 3 | XP_011518168.1 | |||
XM_011519867.1 | 3799 | UTR 3 | XP_011518169.1 | |||
XM_011519868.2 | 3799 | UTR 3 | XP_011518170.1 | |||
XM_017017149.1 | 3799 | UTR 3 | XP_016872638.1 | |||
XM_017017150.1 | 3799 | UTR 3 | XP_016872639.1 | |||
XM_017017151.1 | 3799 | UTR 3 | XP_016872640.1 | |||
XM_017017152.1 | 3799 | UTR 3 | XP_016872641.1 | |||
XM_017017153.1 | 3799 | UTR 3 | XP_016872642.1 | |||
XM_017017154.1 | 3799 | UTR 3 | XP_016872643.1 |
SAA1 - serum amyloid A1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |