Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MIR5006 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU) - Not Available | ||||||
EAS
|
African American
|
YRI (Yoruba) - Not Available | ||||||
SAS
|
Japanese
|
CHB (Han Chinese) - Not Available | ||||||
AFR
|
Chinese
|
JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
MIR5006 - microRNA 5006 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
VWA8 - von Willebrand factor A domain containing 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001009814.1 | Intron | NP_001009814.1 | ||||
NM_015058.1 | Intron | NP_055873.1 | ||||
XM_006719791.3 | Intron | XP_006719854.1 | ||||
XM_011535007.2 | Intron | XP_011533309.1 | ||||
XM_017020469.1 | Intron | XP_016875958.1 | ||||
XM_017020470.1 | Intron | XP_016875959.1 | ||||
XM_017020471.1 | Intron | XP_016875960.1 | ||||
XM_017020472.1 | Intron | XP_016875961.1 | ||||
XM_017020473.1 | Intron | XP_016875962.1 | ||||
XM_017020474.1 | Intron | XP_016875963.1 |