Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACCAGAGCAGCACTCAAATTCATAA[C/G]ATAGGAAATACAAAGGTGCTGCCAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616919 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
FRMPD1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
FRMPD1 - FERM and PDZ domain containing 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TRMT10B - tRNA methyltransferase 10B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286950.1 | Intron | NP_001273879.1 | ||||
NM_001286951.1 | Intron | NP_001273880.1 | ||||
NM_001286952.1 | Intron | NP_001273881.1 | ||||
NM_001286953.1 | Intron | NP_001273882.1 | ||||
NM_001286954.1 | Intron | NP_001273883.1 | ||||
NM_144964.3 | Intron | NP_659401.2 | ||||
XM_005251375.2 | Intron | XP_005251432.1 | ||||
XM_005251379.2 | Intron | XP_005251436.1 | ||||
XM_011517735.2 | Intron | XP_011516037.1 | ||||
XM_011517736.2 | Intron | XP_011516038.1 | ||||
XM_011517738.2 | Intron | XP_011516040.1 | ||||
XM_011517739.2 | Intron | XP_011516041.1 | ||||
XM_011517742.1 | Intron | XP_011516044.1 | ||||
XM_011517743.2 | Intron | XP_011516045.1 | ||||
XM_017014313.1 | Intron | XP_016869802.1 |
Set Membership: |
HapMap |