Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CATTTCACCAAAGGCTGGATGGAGA[A/G]GGGGTGGTGTGTCCAGTGCTAGCCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 601895 | ||||||||||||||||||||
Literature Links: |
MIR4479 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MIR4479 - microRNA 4479 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TRAF2 - TNF receptor associated factor 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_021138.3 | Intron | NP_066961.2 | ||||
XM_011518973.2 | Intron | XP_011517275.1 | ||||
XM_011518974.2 | Intron | XP_011517276.1 | ||||
XM_011518975.2 | Intron | XP_011517277.1 | ||||
XM_011518976.2 | Intron | XP_011517278.1 | ||||
XM_011518977.2 | Intron | XP_011517279.1 | ||||
XM_011518978.2 | Intron | XP_011517280.1 | ||||
XM_017015094.1 | Intron | XP_016870583.1 | ||||
XM_017015095.1 | Intron | XP_016870584.1 |