Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGACAGGAGCTTTGACCTTGACTCT[A/T]CAGGTCCAAGTTGGGTCCCAGCCCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605564 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CIB2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
CIB2 - calcium and integrin binding family member 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001271888.1 | Intron | NP_001258817.1 | ||||
NM_001271889.1 | Intron | NP_001258818.1 | ||||
NM_001301224.1 | Intron | NP_001288153.1 | ||||
NM_006383.3 | Intron | NP_006374.1 | ||||
XM_005254126.3 | Intron | XP_005254183.1 | ||||
XM_006720374.2 | Intron | XP_006720437.1 | ||||
XM_011521161.2 | Intron | XP_011519463.1 |
SH2D7 - SH2 domain containing 7 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |