Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAATGGACTTTGTGAGGTTCCATGT[A/G]TTTGGTCAGTCATGAGAGGAAGAAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF417 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Japanese
|
CHB (Han Chinese) - Not Available | ||||||
AFR
|
Chinese
|
JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
ZNF417 - zinc finger protein 417 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001297734.1 | 3620 | Intron | NP_001284663.1 | |||
NM_152475.2 | 3620 | Intron | NP_689688.2 | |||
XM_011526473.2 | 3620 | UTR 3 | XP_011524775.1 | |||
XM_017026315.1 | 3620 | Intron | XP_016881804.1 |