Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCAGTTTAGGCTAAGGCTGCAAAGC[A/G]GGGACCTACTGAAGATGAATTAGTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609211 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC101927044 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LOC101927044 - uncharacterized LOC101927044 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LOC104968399 - uncharacterized LOC104968399 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MYL12A - myosin light chain 12A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001303047.1 | Intron | NP_001289976.1 | ||||
NM_001303048.1 | Intron | NP_001289977.1 | ||||
NM_001303049.1 | Intron | NP_001289978.1 | ||||
NM_006471.3 | Intron | NP_006462.1 |
MYL12B - myosin light chain 12B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |