Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAAAAAGACAACATGAAGTACAAAA[A/G]GTTACCTTGATTAAACACAAAATCT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
RUFY4 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
RUFY4 - RUN and FYVE domain containing 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_198483.3 | Intron | NP_940885.2 | ||||
XM_011511014.2 | Intron | XP_011509316.1 | ||||
XM_017003895.1 | Intron | XP_016859384.1 | ||||
XM_017003896.1 | Intron | XP_016859385.1 | ||||
XM_017003897.1 | Intron | XP_016859386.1 |
Set Membership: |
HapMap |