Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608435 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MRAS PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
MRAS - muscle RAS oncogene homolog | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001085049.2 | Intron | NP_001078518.1 | ||||
NM_001252090.1 | Intron | NP_001239019.1 | ||||
NM_001252091.1 | Intron | NP_001239020.1 | ||||
NM_001252092.1 | Intron | NP_001239021.1 | ||||
NM_001252093.1 | Intron | NP_001239022.1 | ||||
NM_012219.4 | Intron | NP_036351.3 | ||||
XM_005247228.1 | Intron | XP_005247285.1 | ||||
XM_005247229.2 | Intron | XP_005247286.1 | ||||
XM_017005887.1 | Intron | XP_016861376.1 |
Set Membership: |
HapMap |