Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608708 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
BOC PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
BOC - BOC cell adhesion associated, oncogene regulated | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001301861.1 | Intron | NP_001288790.1 | ||||
NM_001301867.1 | Intron | NP_001288796.1 | ||||
NM_033254.3 | Intron | NP_150279.1 | ||||
XM_005247891.2 | Intron | XP_005247948.1 | ||||
XM_005247892.2 | Intron | XP_005247949.1 | ||||
XM_005247897.2 | Intron | XP_005247954.1 | ||||
XM_011513305.2 | Intron | XP_011511607.1 | ||||
XM_017007441.1 | Intron | XP_016862930.1 | ||||
XM_017007442.1 | Intron | XP_016862931.1 | ||||
XM_017007443.1 | Intron | XP_016862932.1 | ||||
XM_017007444.1 | Intron | XP_016862933.1 | ||||
XM_017007445.1 | Intron | XP_016862934.1 | ||||
XM_017007446.1 | Intron | XP_016862935.1 | ||||
XM_017007447.1 | Intron | XP_016862936.1 | ||||
XM_017007448.1 | Intron | XP_016862937.1 | ||||
XM_017007449.1 | Intron | XP_016862938.1 | ||||
XM_017007450.1 | Intron | XP_016862939.1 | ||||
XM_017007451.1 | Intron | XP_016862940.1 | ||||
XM_017007452.1 | Intron | XP_016862941.1 | ||||
XM_017007453.1 | Intron | XP_016862942.1 | ||||
XM_017007454.1 | Intron | XP_016862943.1 | ||||
XM_017007455.1 | Intron | XP_016862944.1 |