Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605905 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
DCUN1D1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
DCUN1D1 - defective in cullin neddylation 1 domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001308101.1 | 5345 | UTR 3 | NP_001295030.1 | |||
NM_020640.3 | 5345 | UTR 3 | NP_065691.2 | |||
XM_011512912.1 | 5345 | Intron | XP_011511214.1 | |||
XM_011512914.1 | 5345 | Intron | XP_011511216.1 | |||
XM_011512915.2 | 5345 | Intron | XP_011511217.1 | |||
XM_011512916.2 | 5345 | Intron | XP_011511218.1 | |||
XM_017006633.1 | 5345 | Intron | XP_016862122.1 | |||
XM_017006634.1 | 5345 | Intron | XP_016862123.1 | |||
XM_017006635.1 | 5345 | Intron | XP_016862124.1 |