Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 137140 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
GABRA2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
GABRA2 - gamma-aminobutyric acid type A receptor alpha2 subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_005248080.3 | 1780 | UTR 3 | XP_005248137.1 | |||
XM_011513675.1 | 1780 | UTR 3 | XP_011511977.1 | |||
XM_011513676.1 | 1780 | UTR 3 | XP_011511978.1 | |||
XM_017007982.1 | 1780 | UTR 3 | XP_016863471.1 | |||
XM_017007983.1 | 1780 | UTR 3 | XP_016863472.1 |
Set Membership: |
HapMap Validated |