Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 607566 | ||||||||||||||||||||
Literature Links: |
EPM2A PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
EPM2A - epilepsy, progressive myoclonus type 2A, Lafora disease (laforin) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001018041.1 | Intron | NP_001018051.1 | ||||
NM_005670.3 | Intron | NP_005661.1 | ||||
XM_006715564.3 | Intron | XP_006715627.1 | ||||
XM_011536113.2 | Intron | XP_011534415.1 | ||||
XM_011536116.1 | Intron | XP_011534418.1 | ||||
XM_017011300.1 | Intron | XP_016866789.1 | ||||
XM_017011301.1 | Intron | XP_016866790.1 | ||||
XM_017011302.1 | Intron | XP_016866791.1 |