Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 138244 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
GRIK2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
GRIK2 - glutamate ionotropic receptor kainate type subunit 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001166247.1 | Intron | NP_001159719.1 | ||||
NM_021956.4 | Intron | NP_068775.1 | ||||
NM_175768.3 | Intron | NP_786944.1 | ||||
XM_005266945.2 | Intron | XP_005267002.1 | ||||
XM_005266946.3 | Intron | XP_005267003.1 | ||||
XM_011535777.2 | Intron | XP_011534079.1 | ||||
XM_011535778.2 | Intron | XP_011534080.1 | ||||
XM_017010781.1 | Intron | XP_016866270.1 | ||||
XM_017010782.1 | Intron | XP_016866271.1 |
Set Membership: |
HapMap |