Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607638 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
DCTD PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
DCTD - dCMP deaminase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001012732.1 | 1843 | UTR 3 | NP_001012750.1 | |||
NM_001921.2 | 1843 | UTR 3 | NP_001912.2 | |||
XM_005262778.3 | 1843 | UTR 3 | XP_005262835.1 | |||
XM_005262779.3 | 1843 | UTR 3 | XP_005262836.1 | |||
XM_005262780.3 | 1843 | UTR 3 | XP_005262837.1 | |||
XM_005262781.4 | 1843 | UTR 3 | XP_005262838.1 | |||
XM_005262782.3 | 1843 | UTR 3 | XP_005262839.1 | |||
XM_006714115.3 | 1843 | UTR 3 | XP_006714178.1 | |||
XM_006714116.3 | 1843 | UTR 3 | XP_006714179.1 | |||
XM_011531674.2 | 1843 | UTR 3 | XP_011529976.1 | |||
XM_011531675.2 | 1843 | UTR 3 | XP_011529977.1 | |||
XM_011531676.2 | 1843 | UTR 3 | XP_011529978.1 | |||
XM_017007816.1 | 1843 | UTR 3 | XP_016863305.1 | |||
XM_017007817.1 | 1843 | UTR 3 | XP_016863306.1 | |||
XM_017007818.1 | 1843 | UTR 3 | XP_016863307.1 | |||
XM_017007819.1 | 1843 | UTR 3 | XP_016863308.1 | |||
XM_017007820.1 | 1843 | UTR 3 | XP_016863309.1 | |||
XM_017007821.1 | 1843 | UTR 3 | XP_016863310.1 | |||
XM_017007822.1 | 1843 | UTR 3 | XP_016863311.1 | |||
XM_017007823.1 | 1843 | UTR 3 | XP_016863312.1 | |||
XM_017007824.1 | 1843 | UTR 3 | XP_016863313.1 |
Set Membership: |
HapMap |