Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAATTGCTTTCTGCCTATGTATGTA[C/T]GCCTGAAACATTTGCATGAGAGCTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612758 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
TAPT1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
TAPT1 - transmembrane anterior posterior transformation 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_153365.2 | 4329 | UTR 3 | NP_699196.2 | |||
XM_005248139.2 | 4329 | UTR 3 | XP_005248196.1 | |||
XM_005248140.3 | 4329 | UTR 3 | XP_005248197.1 | |||
XM_011513812.2 | 4329 | UTR 3 | XP_011512114.1 | |||
XM_011513815.2 | 4329 | UTR 3 | XP_011512117.1 | |||
XM_011513816.2 | 4329 | UTR 3 | XP_011512118.1 | |||
XM_011513817.2 | 4329 | UTR 3 | XP_011512119.1 | |||
XM_017007876.1 | 4329 | UTR 3 | XP_016863365.1 |